Assay Details
Gene Symbol: Olfr395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147005.1 | NP_667216.1 | 1 | 48 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450039 | NM_147005.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450039 | NM_147005.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162933 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450039 | NM_147005.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450039 | NM_147005.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450039 | NM_147005.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450039 | NM_147005.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918416 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918438 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918443 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918437 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918431 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918439 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918414 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918417 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918435 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918420 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918441 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918412 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918434 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918444 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918409 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918432 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918428 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918413 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918418 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162933 | - | - |
Whole Mouse Genome G4122A | A_51_P450039 | NM_147005.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P450039 | NM_147005.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135440 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162933 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09634672 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09634662 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09360965 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09634674 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09360973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918444 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918410 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918422 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918441 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918414 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918402 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918405 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918407 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918432 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918401 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09634650 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918444 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09360960 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918443 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918438 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918396 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918418 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918412 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918437 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918395 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918408 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918434 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918406 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918404 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918431 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918420 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918439 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918416 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918399 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27918428 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10388295 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10388294 | - | - |
Affymetrix Mouse Clariom S Array | PSR1100029375.mm.1 | NM_147005.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100029375.mm.1 | NM_147005.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100029375.mm.1 | TC1100003182.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 476023 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 102480072 | - | - |
MouseWG-6 v2 Expression BeadChip | 6620324 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6620324 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 102480072 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006620324 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230896 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1217690 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1230896 | - | - |
mouse-6 v1.1 (Illumina) | scl0259007.1_204-S | - | - |
mouse-6 v1.1 (Illumina) | GI_22128932-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102480072 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6620324 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0259007.1_204-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6620324 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102480072 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1230896 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230896 | - | - |
