Assay Details
| Gene Symbol: Gbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172777.3 | NP_766365.1 | 10 - 11 | 1758 | 147 | |
| GenBank mRNA | AK088861.1 | - | 10 - 11 | 1744 | 147 | |
| AK165231.1 | - | 2405 | 147 | |||
| BC057170.1 | - | 1747 | 4978410 | 147 | ||
| DQ985742.1 | - | 1742 | 147 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323180 | DQ985742.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323180 | AK088861.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323180 | AK165231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323180 | NM_172777.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323180 | DQ985742.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323180 | AK088861.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323180 | AK165231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323180 | NM_172777.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292763 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385307 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62068 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323180 | DQ985742.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323180 | AK088861.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323180 | AK165231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323180 | NM_172777.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323180 | DQ985742.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323180 | AK088861.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323180 | AK165231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323180 | NM_172777.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954945 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01178587 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05681775 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05682135 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149481 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107817 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05681914 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01178564 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05682003 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01178585 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05681826 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01178625 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174697 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05681815 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05681987 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05682179 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05682050 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05682164 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01178613 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01178590 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05681802 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05682079 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05681792 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05682103 | - | - |
| Whole Mouse Genome G4122A | A_51_P323180 | DQ985742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323180 | AK088861.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323180 | AK165231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323180 | NM_172777.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323180 | DQ985742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323180 | AK088861.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323180 | AK165231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323180 | NM_172777.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P344949 | DQ985742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P344949 | AK088861.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P344949 | AK165231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P344949 | BC057170.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P344949 | CA535632.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05682079 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149481 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05681802 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01178590 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107817 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05681792 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01178625 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05682135 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01178587 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01178585 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174697 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05682003 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04769867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04472157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04547238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04769867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04705655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23954252 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10532009 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532014 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532005 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532000 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531986 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531984 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531985 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532016 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532017 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531982 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532003 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532018 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532006 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531983 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10531980 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500025582.mm.1 | TC0500003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025582.mm.1 | NM_172777.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025582.mm.1 | NM_172777.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025583.mm.1 | TC0500003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025583.mm.1 | NM_172777.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025583.mm.1 | NM_172777.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025585.mm.1 | TC0500003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025585.mm.1 | NM_172777.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025585.mm.1 | NM_172777.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025572.mm.1 | TC0500003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025572.mm.1 | NM_172777.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025572.mm.1 | NM_172777.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025576.mm.1 | TC0500003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025576.mm.1 | NM_172777.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025576.mm.1 | NM_172777.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025578.mm.1 | TC0500003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025578.mm.1 | NM_172777.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025578.mm.1 | NM_172777.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025580.mm.1 | TC0500003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025580.mm.1 | NM_172777.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025580.mm.1 | NM_172777.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025577.mm.1 | TC0500003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025577.mm.1 | NM_172777.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025577.mm.1 | NM_172777.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025575.mm.1 | TC0500003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025575.mm.1 | NM_172777.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025575.mm.1 | NM_172777.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025579.mm.1 | TC0500003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025579.mm.1 | NM_172777.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025579.mm.1 | NM_172777.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025581.mm.1 | TC0500003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025581.mm.1 | NM_172777.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025581.mm.1 | NM_172777.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025584.mm.1 | TC0500003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025584.mm.1 | NM_172777.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025584.mm.1 | NM_172777.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500033832.mm.1 | TC0500003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500033832.mm.1 | NM_172777.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500033832.mm.1 | NM_172777.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 365875 | - | - |
| ABI Mouse Genome Survey Microarray | 409840 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 780204 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3290594 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 780204 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000780204 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003290594 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2603001 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2641863 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2603001 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2641863 | - | - |
| mouse-6 v1.1 (Illumina) | scl00236573.2_324-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00236573.1_47-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 780204 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3290594 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00236573.2_324-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00236573.1_47-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 780204 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3290594 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2603001 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2641863 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2603001 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2641863 | - | - |

Gbp9
Genomic Map