Assay Details
| Gene Symbol: Csf2rb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287389.1 | NP_001274318.1 | 7 - 8 | 1125 | 96 | |
| NM_007781.3 | NP_031807.1 | 9 - 10 | 1452 | 96 | ||
| GenBank mRNA | AK154650.1 | - | 9 - 10 | 1375 | 96 | |
| AK155216.1 | - | 9 - 10 | 1378 | 96 | ||
| AK155511.1 | - | 9 - 10 | 1354 | 96 | ||
| AK155626.1 | - | 7 - 8 | 1027 | 96 | ||
| AK155675.1 | - | 9 - 10 | 1354 | 96 | ||
| AK170028.1 | - | 9 - 10 | 1354 | 96 | ||
| M29855.1 | - | 9 - 10 | 1334 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146753 | M29855.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146753 | NM_007781.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146753 | M29855.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146753 | NM_007781.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034663 | M29855.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034663 | AK155626.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034663 | NM_007781.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149742 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286442 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883297 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808828 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 433409 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486036 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 58843 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 383484 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534385 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 733631 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425910 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 142159 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111969 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105528 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154362 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034663 | M29855.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034663 | AK155626.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034663 | NM_007781.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146753 | M29855.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146753 | NM_007781.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146753 | M29855.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146753 | NM_007781.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146753 | M29855.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146753 | NM_007781.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146753 | M29855.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146753 | NM_007781.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146753 | M29855.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146753 | NM_007781.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146753 | M29855.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146753 | NM_007781.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116410 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12423641 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11841860 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11841864 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11841862 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11841851 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11841861 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10430308 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430310 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430304 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430305 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430307 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430309 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430303 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430306 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449360_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430302 | - | - |
| Mouse Genome 430 2.0 Array | 1449360_at | - | - |
| Mouse Genome 430 2.0 Array | 1449360_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12984_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12984_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500014283.mm.1 | NM_001287389.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014283.mm.1 | NM_007781.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014283.mm.1 | TC1500001828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014287.mm.1 | NM_007781.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014287.mm.1 | TC1500001828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014294.mm.1 | NM_001287389.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014294.mm.1 | NM_007781.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014294.mm.1 | TC1500001828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014285.mm.1 | NM_001287389.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014285.mm.1 | NM_007781.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014285.mm.1 | TC1500001828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014286.mm.1 | NM_001287389.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014286.mm.1 | NM_007781.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014286.mm.1 | TC1500001828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014291.mm.1 | NM_001287389.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014291.mm.1 | NM_007781.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014291.mm.1 | TC1500001828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014290.mm.1 | NM_001287389.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014290.mm.1 | NM_007781.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014290.mm.1 | TC1500001828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014293.mm.1 | NM_001287389.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014293.mm.1 | NM_007781.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014293.mm.1 | TC1500001828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014288.mm.1 | NM_007781.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014288.mm.1 | TC1500001828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014289.mm.1 | NM_001287389.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014289.mm.1 | NM_007781.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014289.mm.1 | TC1500001828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014284.mm.1 | NM_001287389.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014284.mm.1 | NM_007781.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014284.mm.1 | TC1500001828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014292.mm.1 | NM_001287389.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014292.mm.1 | NM_007781.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014292.mm.1 | TC1500001828.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449360_PM_at | - | - |
| Mouse Expression Set 430 | 1450200_PM_s_at | - | - |
| Mouse Expression Set 430 | 1449360_at | - | - |
| Mouse Expression Set 430 | 1450200_s_at | - | - |
| Murine Genome U74v2 Set | 103210_at | - | - |
| Murine Genome U74v2 Set | 103210_PM_at | - | - |
| Murine Genome U74v2 Set | 161544_PM_r_at | - | - |
| Murine Genome U74v2 Set | 161544_r_at | - | - |
| Murine Genome U74v2 Set | 94748_g_at | - | - |
| Murine Genome U74v2 Set | 94748_PM_g_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449360_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449360_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450200_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450200_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449360_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449360_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 486066 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3120546 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3120546 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003120546 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2750047 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2750047 | - | - |
| mouse-6 v1.1 (Illumina) | scl46994.14_16-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3120546 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46994.14_16-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3120546 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2750047 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2750047 | - | - |

Csf2rb2
Genomic Map