Assay Details
| Gene Symbol: Mesp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008589.2 | NP_032615.2 | 1 - 2 | 1033 | 140 | |
| GenBank mRNA | AK146740.1 | - | 1 - 2 | 513 | 140 | |
| BC145652.1 | - | 1 - 2 | 863 | 40130955 | 140 | |
| BC145654.1 | - | 1 - 2 | 863 | 40130957 | 140 | |
| U71125.1 | - | 1 - 2 | 1038 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
somitogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
Notch signaling pathway
multicellular organism development
mesodermal cell migration
signal transduction involved in regulation of gene expression
somite rostral/caudal axis specification
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein dimerization activity
somitogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
Notch signaling pathway
multicellular organism development
mesodermal cell migration
signal transduction involved in regulation of gene expression
somite rostral/caudal axis specification
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein dimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25263317 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25263319 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25263318 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317433 | AK146740.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317433 | U71125.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317433 | NM_008589.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317433 | AK146740.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317433 | U71125.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317433 | NM_008589.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352451 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 120444 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168917 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317433 | AK146740.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317433 | U71125.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317433 | NM_008589.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317433 | AK146740.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317433 | U71125.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317433 | NM_008589.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317433 | AK146740.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317433 | U71125.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317433 | NM_008589.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317433 | AK146740.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317433 | U71125.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317433 | NM_008589.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25263333 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168917 | - | - |
| Whole Mouse Genome G4122A | A_51_P317433 | AK146740.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317433 | U71125.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317433 | NM_008589.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317433 | AK146740.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317433 | U71125.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317433 | NM_008589.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118498 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135171 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168917 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06579867 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06579863 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06579869 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06398087 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06579870 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06579850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25263319 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25263317 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25263318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25263273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06193766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05895582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06621923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05990195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06621925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05920155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06579870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06579863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06217120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06153205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06011689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05895583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06193769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06217119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06293058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06551773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06011688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06579869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25263348 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10554368 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554369 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419629_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10554367 | - | - |
| Mouse Genome 430 2.0 Array | 1419629_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419629_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17293_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17293_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700009989.mm.1 | TC0700001135.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700009989.mm.1 | NM_008589.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009989.mm.1 | NM_008589.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009992.mm.1 | TC0700001135.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700009992.mm.1 | NM_008589.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009992.mm.1 | NM_008589.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009990.mm.1 | TC0700001135.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700009990.mm.1 | NM_008589.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009990.mm.1 | NM_008589.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009991.mm.1 | TC0700001135.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700009991.mm.1 | NM_008589.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009991.mm.1 | NM_008589.2 | Refseq |
| Mouse Expression Set 430 | 1419629_PM_at | - | - |
| Mouse Expression Set 430 | 1419629_at | - | - |
| Murine Genome U74v2 Set | 97092_PM_at | - | - |
| Murine Genome U74v2 Set | 97092_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419629_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419629_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419629_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419629_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 541607 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5550736 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005550736 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2713960 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2713960 | - | - |
| mouse-6 v1.1 (Illumina) | scl32487.2.1_102-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5550736 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32487.2.1_102-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550736 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2713960 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2713960 | - | - |

Mesp2
Genomic Map