Assay Details
| Gene Symbol: Prm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008933.1 | NP_032959.1 | 1 - 2 | 359 | 153 | |
| GenBank mRNA | AK005729.1 | - | 1 - 2 | 368 | 153 | |
| BC049612.1 | - | 1 - 2 | 364 | 6774072 | 153 | |
| M16456.1 | - | 1 - 2 | 262 | 153 | ||
| M27501.1 | - | 1 - 2 | 262 | 153 | ||
| X14004.1 | - | 1 - 2 | 359 | 153 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P138705 | NM_008933.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P138705 | AK005729.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P138705 | BC049612.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P138705 | M16456.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P138705 | M27501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P138705 | NM_008933.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P138705 | AK005729.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P138705 | BC049612.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P138705 | M16456.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P138705 | M27501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P138705 | NM_008933.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P138705 | AK005729.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P138705 | BC049612.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P138705 | M16456.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P138705 | M27501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P138705 | NM_008933.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P138705 | AK005729.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P138705 | BC049612.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P138705 | M16456.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P138705 | M27501.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P138705 | NM_008933.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P138705 | AK005729.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P138705 | BC049612.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P138705 | M16456.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P138705 | M27501.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P138705 | NM_008933.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P138705 | AK005729.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P138705 | BC049612.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P138705 | M16456.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P138705 | M27501.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544520 | - | - |
| Whole Mouse Genome G4122A | A_51_P138705 | NM_008933.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P138705 | AK005729.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P138705 | BC049612.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P138705 | M16456.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P138705 | M27501.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P138705 | NM_008933.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P138705 | AK005729.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P138705 | BC049612.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P138705 | M16456.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P138705 | M27501.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544520 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10437682 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437683 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437681 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448105_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10437680 | - | - |
| Mouse Genome 430 2.0 Array | 1448105_at | - | - |
| Mouse Genome 430 2.0 Array | 1448105_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19119_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19119_at | - | - |
| Mouse Expression Set 430 | 1448105_at | - | - |
| Mouse Expression Set 430 | 1448105_PM_at | - | - |
| Murine Genome U74v2 Set | 160528_PM_at | - | - |
| Murine Genome U74v2 Set | 160528_at | - | - |
| Murine Genome U74v2 Set | 170868_at | - | - |
| Murine Genome U74v2 Set | 170873_i_at | - | - |
| Murine Genome U74v2 Set | 171105_PM_at | - | - |
| Murine Genome U74v2 Set | 170868_PM_at | - | - |
| Murine Genome U74v2 Set | 170873_PM_i_at | - | - |
| Murine Genome U74v2 Set | 171105_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448105_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448105_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448105_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448105_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 529019 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3060368 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3060368 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003060368 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2624909 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2624909 | - | - |
| mouse-6 v1.1 (Illumina) | scl019119.2_47-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3060368 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl019119.2_47-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3060368 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl019119.2_47-S | - | - |

Prm2
Genomic Map