Assay Details
| Gene Symbol: Zfp106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U59739.1 | - | 2 - 3 | 3469 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp106| Gene Symbol: Zfp106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U59739.1 | - | 2 - 3 | 3469 | 124 | |
Set Membership: |
|