Assay Details
| Gene Symbol: Lyz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013590.4 | NP_038618.1 | 1 - 2 | 163 | 129 | |
| GenBank mRNA | BC061129.1 | - | 1 - 2 | 163 | 30296274 | 129 |
| X51547.1 | - | 1 - 2 | 133 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lyz1
Genomic Map