Assay Details
| Gene Symbol: Krt31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010659.2 | NP_034789.2 | 2 - 3 | 499 | 143 | |
| GenBank mRNA | M27734.1 | - | 2 - 3 | 499 | 143 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2140042 | NM_010659.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2140042 | M27734.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919046 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 13171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 938428 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159412 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163547 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2140042 | NM_010659.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2140042 | M27734.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P304200 | NM_010659.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P304200 | M27734.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28065699 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163547 | - | - |
| Whole Mouse Genome G4122A | A_51_P304200 | NM_010659.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P304200 | M27734.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124684 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163547 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08984731 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09431575 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09431559 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09260663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28065737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09716657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09291383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09544522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09006943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09256117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09539599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09504108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28065746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08984729 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10390980 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390984 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390983 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390982 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421589_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10390979 | - | - |
| Mouse Genome 430 2.0 Array | 1421589_at | - | - |
| Mouse Genome 430 2.0 Array | 1421589_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16660_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16660_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100034405.mm.1 | NM_010659.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034405.mm.1 | NM_010659.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034405.mm.1 | TC1100003751.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100034403.mm.1 | NM_010659.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034403.mm.1 | NM_010659.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034403.mm.1 | TC1100003751.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100034406.mm.1 | NM_010659.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034406.mm.1 | NM_010659.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034406.mm.1 | TC1100003751.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100034404.mm.1 | NM_010659.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034404.mm.1 | NM_010659.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034404.mm.1 | TC1100003751.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100034407.mm.1 | NM_010659.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034407.mm.1 | NM_010659.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034407.mm.1 | TC1100003751.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100034408.mm.1 | NM_010659.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034408.mm.1 | NM_010659.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034408.mm.1 | TC1100003751.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421589_PM_at | - | - |
| Mouse Expression Set 430 | 1421589_at | - | - |
| Murine Genome U74v2 Set | 103558_PM_at | - | - |
| Murine Genome U74v2 Set | 103558_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421589_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421589_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421589_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421589_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6450142 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006450142 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2622236 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2622236 | - | - |
| mouse-6 v1.1 (Illumina) | scl39577.7.1_81-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6450142 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39577.7.1_81-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6450142 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2622235 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2622235 | - | - |

Krt31
Genomic Map