Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0Y00000088.mm.1 |
NM_012011.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0Y00000088.mm.1 |
NM_012011.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0Y00000088.mm.1 |
TC0Y00000006.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0Y00000089.mm.1 |
NM_012011.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0Y00000089.mm.1 |
NM_012011.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0Y00000089.mm.1 |
TC0Y00000006.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0Y00000090.mm.1 |
NM_012011.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0Y00000090.mm.1 |
NM_012011.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0Y00000090.mm.1 |
TC0Y00000006.mm.1 |
MTA TC |