Assay Details
| Gene Symbol: G6pd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019468.2 | NP_062341.2 | 1 | 1181 | 65 | |
| GenBank mRNA | BC120827.1 | - | 1 | 1181 | 40129750 | 65 |
| BC137684.1 | - | 1 | 1181 | 8860703 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353735 | BC120827.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353735 | BC137684.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353735 | BC120827.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353735 | BC137684.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210231 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 629977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20725 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179107 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788368 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 774239 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144239 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 161077 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137273 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 70390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 97392 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110110 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140818 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353735 | BC120827.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353735 | BC137684.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353735 | BC120827.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353735 | BC137684.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353735 | BC120827.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353735 | BC137684.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353735 | BC120827.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353735 | BC137684.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23738028 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140818 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110110 | - | - |
| Whole Mouse Genome G4122A | A_51_P353735 | BC120827.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353735 | BC137684.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353735 | BC120827.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353735 | BC137684.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119009 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110110 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140818 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04835244 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04835245 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04835229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23737998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23737999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23737992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23737995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23738023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04954229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738062 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04835241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23737990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04818815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23737991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04814298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04136768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04558089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738017 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23738023 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10521985 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422326_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10521984 | - | - |
| Mouse Genome 430 2.0 Array | 1422327_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1422327_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1422326_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422326_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14380_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14380_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500005153.mm.1 | TC0500000580.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500005153.mm.1 | NM_019468.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500005153.mm.1 | NM_019468.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500005154.mm.1 | TC0500000580.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500005154.mm.1 | NM_019468.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500005154.mm.1 | NM_019468.2 | Refseq |
| Mouse Expression Set 430 | 1422326_PM_at | - | - |
| Mouse Expression Set 430 | 1422326_at | - | - |
| Mouse Expression Set 430 | 1422327_s_at | - | - |
| Mouse Expression Set 430 | 1422327_PM_s_at | - | - |
| Murine Genome U74v2 Set | 101293_PM_at | - | - |
| Murine Genome U74v2 Set | 101294_g_at | - | - |
| Murine Genome U74v2 Set | 101293_at | - | - |
| Murine Genome U74v2 Set | 101294_PM_g_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422327_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422326_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422326_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422327_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422326_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422326_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 589780 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6900014 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006900014 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231565 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231565 | - | - |
| mouse-6 v1.1 (Illumina) | scl014380.1_82-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6900014 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl014380.1_82-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900014 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2617607 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2943790 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2943790 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2617607 | - | - |

G6pd2
Genomic Map