Assay Details
| Gene Symbol: Zfp235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019941.2 | NP_064325.2 | 3 - 4 | 509 | 147 | |
| GenBank mRNA | AB221682.1 | - | 1 - 2 | 141 | 147 | |
| AF167321.1 | - | 3 - 4 | 470 | 147 | ||
| BC099965.1 | - | 3 - 4 | 430 | 30084081 | 147 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25003662 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25003661 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038697 | BC099965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038697 | AF167321.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038697 | NM_019941.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 707728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 133357 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204335 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649971 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371048 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 231489 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 60869 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122688 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038697 | BC099965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038697 | AF167321.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038697 | NM_019941.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P220414 | BC099965.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P220414 | AF167321.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P220414 | NM_019941.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003677 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122688 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01484973 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06140261 | - | - |
| Whole Mouse Genome G4122A | A_51_P220414 | BC099965.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P220414 | AF167321.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P220414 | NM_019941.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132691 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122688 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06022716 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06022714 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06022717 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25003661 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25003662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06251748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05966383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06471544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06094699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06251745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05966387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05966398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06022717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05966386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05966394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05971720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05966389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05966399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06124607 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10550848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550853 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550850 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550849 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550854 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550852 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429988_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449329_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10550847 | - | - |
| Mouse Genome 430 2.0 Array | 1429988_at | - | - |
| Mouse Genome 430 2.0 Array | 1429988_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449329_at | - | - |
| Mouse Genome 430 2.0 Array | 1449329_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56525_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56525_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700002680.mm.1 | TC0700000370.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002680.mm.1 | NM_019941.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002680.mm.1 | NM_019941.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002684.mm.1 | TC0700000370.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002684.mm.1 | NM_019941.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002684.mm.1 | NM_019941.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002693.mm.1 | TC0700000370.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002693.mm.1 | NM_019941.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002693.mm.1 | NM_019941.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002682.mm.1 | TC0700000370.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002682.mm.1 | NM_019941.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002682.mm.1 | NM_019941.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002692.mm.1 | TC0700000370.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002692.mm.1 | NM_019941.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002692.mm.1 | NM_019941.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002690.mm.1 | TC0700000370.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002690.mm.1 | NM_019941.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002690.mm.1 | NM_019941.2 | Refseq |
| Mouse Expression Set 430 | 1449329_at | - | - |
| Mouse Expression Set 430 | 1449329_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449329_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449329_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449329_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449329_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429988_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429988_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 826626 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1690066 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1690066 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001690066 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2466926 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2466926 | - | - |
| mouse-6 v1.1 (Illumina) | scl32973.3.1_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1690066 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32973.3.1_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1690066 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2466926 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2466926 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32973.3.1_23-S | - | - |

Zfp235
Genomic Map