Assay Details
| Gene Symbol: Drd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK032054.1 | - | 1 | 2477 | 152 | |
| AK045456.1 | - | 1 | 2666 | 152 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Drd5
Genomic Map