Assay Details
| Gene Symbol: Trp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290830.1 | NP_001277759.1 | 22 - 23 | 5043 | 76 | |
| NM_013735.4 | NP_038763.3 | 23 - 24 | 5193 | 76 | ||
| GenBank mRNA | AJ414734.1 | - | 22 - 23 | 5036 | 76 | |
| AK029832.1 | - | 1590 | 76 | |||
| AK036324.1 | - | 4 - 5 | 998 | 76 | ||
| AK132323.1 | - | 5 - 6 | 1226 | 76 | ||
| BC035206.1 | - | 11 - 12 | 2405 | 3979746 | 76 | |
| BC079906.1 | - | 22 - 23 | 4977 | 30639375 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp1
Genomic Map