Assay Details
| Gene Symbol: Trp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290830.1 | NP_001277759.1 | 24 - 25 | 5358 | 73 | |
| NM_013735.4 | NP_038763.3 | 25 - 26 | 5508 | 73 | ||
| GenBank mRNA | AJ414734.1 | - | 24 - 25 | 5351 | 73 | |
| AK029832.1 | - | 1905 | 73 | |||
| AK036324.1 | - | 6 - 7 | 1313 | 73 | ||
| AK132323.1 | - | 7 - 8 | 1541 | 73 | ||
| BC035206.1 | - | 13 - 14 | 2720 | 3979746 | 73 | |
| BC079906.1 | - | 24 - 25 | 5292 | 30639375 | 73 | |
| CA327869.1 | - | 2 - 3 | 128 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp1
Genomic Map