Assay Details
| Gene Symbol: Rgs6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282061.1 | NP_001268990.1 | 4 - 5 | 326 | 77 | |
| NM_015812.3 | NP_056627.1 | 4 - 5 | 608 | 77 | ||
| GenBank mRNA | AF061933.1 | - | 1 - 2 | 53 | 77 | |
| AK052527.1 | - | 4 - 5 | 609 | 77 | ||
| AK053087.1 | - | 4 - 5 | 327 | 77 | ||
| AK139323.1 | - | 4 - 5 | 723 | 77 | ||
| BC038616.1 | - | 2 - 3 | 153 | 77 | ||
| BC125029.1 | - | 4 - 5 | 312 | 40062355 | 77 | |
| BC125030.1 | - | 4 - 5 | 312 | 40062356 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rgs6
Genomic Map