Assay Details
| Gene Symbol: Cnppd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026977.2 | NP_081253.1 | 3 - 4 | 506 | 91 | |
| GenBank mRNA | AK029090.1 | - | 3 - 4 | 506 | 91 | |
| AK145962.1 | - | 4 - 5 | 753 | 91 | ||
| AK159252.1 | - | 228 | 91 | |||
| AK169779.1 | - | 3 - 4 | 778 | 91 | ||
| AK187356.1 | - | 2 - 3 | 126 | 91 | ||
| BC013714.1 | - | 2 - 3 | 279 | 91 | ||
| BC028815.1 | - | 3 - 4 | 491 | 5126467 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnppd1
Genomic Map