Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200022014.mm.1 |
NM_001102423.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022014.mm.1 |
NM_001102423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022014.mm.1 |
NM_001102424.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022014.mm.1 |
NM_001102424.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022014.mm.1 |
NM_001102425.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022014.mm.1 |
NM_001102425.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022014.mm.1 |
NM_172675.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022014.mm.1 |
NM_172675.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022014.mm.1 |
TC0200002696.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200022017.mm.1 |
NM_001102423.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022017.mm.1 |
NM_001102423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022017.mm.1 |
NM_172675.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022017.mm.1 |
NM_172675.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022017.mm.1 |
TC0200002696.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200022020.mm.1 |
NM_001102423.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022020.mm.1 |
NM_001102423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022020.mm.1 |
NM_001102424.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022020.mm.1 |
NM_001102424.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022020.mm.1 |
NM_001102425.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022020.mm.1 |
NM_001102425.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022020.mm.1 |
NM_172675.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022020.mm.1 |
NM_172675.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022020.mm.1 |
TC0200002696.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200022022.mm.1 |
NM_001102423.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022022.mm.1 |
NM_001102423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022022.mm.1 |
NM_001102424.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022022.mm.1 |
NM_001102424.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022022.mm.1 |
NM_001102425.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022022.mm.1 |
NM_001102425.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022022.mm.1 |
NM_172675.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022022.mm.1 |
NM_172675.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022022.mm.1 |
TC0200002696.mm.1 |
MTA TC |