| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800002089.mm.1 |
NM_011394.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002089.mm.1 |
NM_011394.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002089.mm.1 |
TC0800000277.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800002086.mm.1 |
NM_011394.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002086.mm.1 |
NM_011394.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002086.mm.1 |
TC0800000277.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800002088.mm.1 |
NM_011394.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002088.mm.1 |
NM_011394.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002088.mm.1 |
TC0800000277.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800002087.mm.1 |
NM_011394.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002087.mm.1 |
NM_011394.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002087.mm.1 |
TC0800000277.mm.1 |
MTA TC |