Assay Details
| Gene Symbol: Slc20a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011394.3 | NP_035524.2 | 3 - 4 | 925 | 69 | |
| GenBank mRNA | AF196476.1 | - | 2 - 3 | 693 | 69 | |
| AK087507.1 | - | 3 - 4 | 70 | |||
| AK157206.1 | - | 3 - 4 | 799 | 69 | ||
| AK170985.1 | - | 3 - 4 | 891 | 69 | ||
| AK177809.1 | - | 2 - 3 | 207 | 69 | ||
| AK181075.1 | - | 2 - 3 | 206 | 69 | ||
| AK208287.1 | - | 2 - 3 | 207 | 69 | ||
| BC046510.1 | - | 3 - 4 | 848 | 4035182 | 69 | |
| BC094562.1 | - | 3 - 4 | 1305 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc20a2
Genomic Map