Assay Details
| Gene Symbol: Slc20a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011394.3 | NP_035524.2 | 7 - 8 | 1424 | 62 | |
| GenBank mRNA | AF196476.1 | - | 6 - 7 | 1192 | 62 | |
| AK087507.1 | - | 7 - 8 | 1249 | 62 | ||
| AK157206.1 | - | 7 - 8 | 1298 | 62 | ||
| AK170985.1 | - | 7 - 8 | 1390 | 62 | ||
| BC046510.1 | - | 7 - 8 | 1347 | 4035182 | 62 | |
| BC094562.1 | - | 7 - 8 | 1804 | 62 | ||
| CF949043.1 | - | 1 - 2 | 215 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc20a2
Genomic Map