Assay Details
| Gene Symbol: Trp73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126330.1 | NP_001119802.1 | 10 - 11 | 1587 | 115 | |
| NM_011642.3 | NP_035772.2 | 12 - 13 | 1877 | 115 | ||
| GenBank mRNA | AB221594.1 | - | 11 - 12 | 1469 | 115 | |
| AK014503.1 | - | 5 - 6 | 566 | 115 | ||
| AK017412.1 | - | 9 - 10 | 1118 | 115 | ||
| BC066045.1 | - | 10 - 11 | 1474 | 6812399 | 115 | |
| BC069182.1 | - | 6 - 7 | 734 | 115 | ||
| Y19234.1 | - | 12 - 13 | 1878 | 115 | ||
| Y19235.1 | - | 10 - 11 | 1581 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp73
Genomic Map