Assay Details
| Gene Symbol: Trp73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011642.3 | NP_035772.2 | 3 - 4 | 570 | 71 | |
| GenBank mRNA | AB221594.1 | - | 2 - 3 | 162 | 71 | |
| BB663805.1 | - | 3 - 4 | 306 | 71 | ||
| Y19234.1 | - | 3 - 4 | 571 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp73
Genomic Map