Assay Details
| Gene Symbol: Trp73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126330.1 | NP_001119802.1 | 7 - 8 | 1168 | 108 | |
| NM_001126331.1 | NP_001119803.1 | 7 - 8 | 1168 | 108 | ||
| NM_011642.3 | NP_035772.2 | 9 - 10 | 1458 | 108 | ||
| GenBank mRNA | AB221594.1 | - | 8 - 9 | 1050 | 108 | |
| AK014503.1 | - | 2 - 3 | 147 | 108 | ||
| AK017412.1 | - | 6 - 7 | 699 | 108 | ||
| AK139633.1 | - | 7 - 8 | 1169 | 108 | ||
| BC066045.1 | - | 7 - 8 | 1055 | 6812399 | 108 | |
| BC069182.1 | - | 3 - 4 | 315 | 108 | ||
| Y19234.1 | - | 9 - 10 | 1459 | 108 | ||
| Y19235.1 | - | 7 - 8 | 1162 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014503.1 transcripts.
Set Membership: |
|

Trp73
Genomic Map