Assay Details
Gene Symbol: Nagpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013796.3 | NP_038824.2 | 9 - 10 | 1367 | 107 | |
NR_027804.1 | 8 - 9 | 1464 | 107 | |||
GenBank mRNA | AK032158.1 | - | 9 - 10 | 1368 | 107 | |
AK087468.1 | - | 9 - 10 | 1370 | 107 | ||
AK138035.1 | - | 9 - 10 | 1360 | 107 | ||
AK162633.1 | - | 9 - 10 | 1350 | 107 | ||
AK168830.1 | - | 8 - 9 | 1452 | 107 | ||
BC019640.1 | - | 8 - 9 | 1429 | 107 | ||
BC039790.1 | - | 9 - 10 | 1359 | 4951805 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30514373 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30514372 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30514374 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30514377 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30514379 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30514378 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370050 | AK162633.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370050 | AK168830.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370050 | AK159629.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370050 | AK138035.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370050 | AK087468.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370050 | AK162633.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370050 | AK168830.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370050 | AK159629.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370050 | AK138035.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370050 | AK087468.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 856321 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61523 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 956940 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741136 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254872 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 165021 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 148868 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124659 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115112 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128753 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370050 | AK162633.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370050 | AK168830.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370050 | AK159629.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370050 | AK138035.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370050 | AK087468.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370050 | AK162633.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370050 | AK168830.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370050 | AK159629.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370050 | AK138035.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370050 | AK087468.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514376 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514368 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514387 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514395 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514367 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514373 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514380 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514361 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514359 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514385 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514360 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514363 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514375 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514365 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514389 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514404 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514407 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514403 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514370 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514399 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514369 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514382 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514392 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514396 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514394 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30514378 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115112 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128753 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124659 | - | - |
Whole Mouse Genome G4122A | A_51_P370050 | AK162633.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P370050 | AK168830.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P370050 | AK159629.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P370050 | AK138035.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P370050 | AK087468.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P370050 | AK162633.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P370050 | AK168830.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P370050 | AK159629.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P370050 | AK138035.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P370050 | AK087468.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121382 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128753 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124659 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115112 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12608580 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12555616 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12608567 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30514378 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30514373 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30514379 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30514377 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30514374 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30514372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30514359 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514299 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514380 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514394 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514290 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514351 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514368 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12877936 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514339 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514414 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12673195 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514344 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514392 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12608572 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514382 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514333 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514361 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514363 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12963651 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514387 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514408 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514383 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514407 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514356 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12555616 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12608584 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514367 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514349 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514360 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12522740 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12577126 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514404 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514399 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514295 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514340 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514370 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514390 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514347 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12704931 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13041330 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514401 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514373 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12622751 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514410 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12673196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514396 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12815797 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514294 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12963653 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514330 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514359 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12730186 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514343 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13027709 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30514338 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12846738 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10437551 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437544 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437546 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437543 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437547 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437542 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437548 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437549 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437550 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10437545 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1454718_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1458344_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10437541 | - | - |
Mouse Genome 430 2.0 Array | 1454718_at | - | - |
Mouse Genome 430 2.0 Array | 1458344_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1454718_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1458344_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27426_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27426_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1600009248.mm.1 | NM_013796.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600009248.mm.1 | NM_013796.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600009248.mm.1 | NR_027804.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600009248.mm.1 | NR_027804.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600009248.mm.1 | TC1600001155.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1600009247.mm.1 | NM_013796.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600009247.mm.1 | NM_013796.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600009247.mm.1 | NR_027804.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600009247.mm.1 | NR_027804.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600009247.mm.1 | TC1600001155.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1600009249.mm.1 | NM_013796.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600009249.mm.1 | NM_013796.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600009249.mm.1 | NR_027804.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600009249.mm.1 | NR_027804.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600009249.mm.1 | TC1600001155.mm.1 | MTA TC |
Mouse Expression Set 430 | 1454718_at | - | - |
Mouse Expression Set 430 | 1454718_PM_at | - | - |
Mouse Expression Set 430 | 1458344_PM_at | - | - |
Mouse Expression Set 430 | 1458344_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458344_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454718_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454718_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458344_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1770053 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1770053 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001770053 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2654635 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2654635 | - | - |
mouse-6 v1.1 (Illumina) | scl48790.9.1_54-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1770053 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl48790.9.1_54-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1770053 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2654635 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2654635 | - | - |
