Assay Details
| Gene Symbol: Tab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025609.2 | NP_079885.2 | 5 - 6 | 554 | 78 | |
| GenBank mRNA | AB088136.1 | - | 5 - 6 | 554 | 78 | |
| BC027054.1 | - | 5 - 6 | 546 | 78 | ||
| BC041110.1 | - | 5 - 6 | 549 | 78 | ||
| BC054369.1 | - | 5 - 6 | 554 | 5356886 | 78 | |
| BC114637.1 | - | 5 - 6 | 575 | 78 | ||
More Information
Set Membership: |
|

Tab1
Genomic Map