Assay Details
| Gene Symbol: Acbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025988.2 | NP_080264.1 | 6 - 7 | 961 | 116 | |
| GenBank mRNA | AK008243.1 | - | 4 - 5 | 510 | 116 | |
| AK148438.1 | - | 456 | 116 | |||
| BC048371.1 | - | 5 - 6 | 875 | 6396076 | 116 | |
| BI734653.1 | - | 5 - 6 | 495 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083324 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292757 | AK165381.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292757 | BC048371.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292757 | AK079604.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292757 | AK148438.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292757 | NM_025988.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292757 | AK165381.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292757 | BC048371.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292757 | AK079604.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292757 | AK148438.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292757 | NM_025988.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139611 | AK165381.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139611 | BC048371.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139611 | AK079604.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139611 | AK148438.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139611 | NM_025988.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360128 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814092 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727871 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 954803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40947 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 791548 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552634 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458960 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349615 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 78299 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180414 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165322 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292757 | AK165381.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292757 | BC048371.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292757 | AK079604.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292757 | AK148438.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292757 | NM_025988.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292757 | AK165381.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292757 | BC048371.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292757 | AK079604.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292757 | AK148438.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292757 | NM_025988.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139611 | AK165381.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139611 | BC048371.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139611 | AK079604.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139611 | AK148438.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139611 | NM_025988.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P292757 | AK165381.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P292757 | BC048371.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P292757 | AK079604.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P292757 | AK148438.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P292757 | NM_025988.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P292757 | AK165381.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P292757 | BC048371.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P292757 | AK079604.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P292757 | AK148438.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P292757 | NM_025988.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083367 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07471274 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165322 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07471292 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180414 | - | - |
| Whole Mouse Genome G4122A | A_51_P292757 | AK165381.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P292757 | BC048371.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P292757 | AK079604.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P292757 | AK148438.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P292757 | NM_025988.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P292757 | AK165381.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P292757 | BC048371.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P292757 | AK079604.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P292757 | AK148438.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P292757 | NM_025988.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129898 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165322 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180414 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083324 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10381686 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381693 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381689 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381685 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381694 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381688 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381692 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381691 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381684 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381687 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449626_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428271_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10381683 | - | - |
| Mouse Genome 430 2.0 Array | 1428271_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428271_at | - | - |
| Mouse Genome 430 2.0 Array | 1449626_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1449626_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67131_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67131_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100016103.mm.1 | NM_025988.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100016103.mm.1 | NM_025988.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100016103.mm.1 | TC1100001705.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100016104.mm.1 | NM_025988.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100016104.mm.1 | NM_025988.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100016104.mm.1 | TC1100001705.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428271_PM_at | - | - |
| Mouse Expression Set 430 | 1428271_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428271_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1449626_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1449626_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428271_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 916283 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6980079 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6980079 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006980079 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2689540 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2689540 | - | - |
| mouse-6 v1.1 (Illumina) | scl40849.10_252-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6980079 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40849.10_252-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980079 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3071515 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2689540 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3150296 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3150296 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2689540 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3071515 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40849.10_252-S | - | - |

Acbd4
Genomic Map