Assay Details
| Gene Symbol: Tbc1d23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026254.2 | NP_080530.2 | 3 - 4 | 373 | 89 | |
| GenBank mRNA | AK155142.1 | - | 3 - 4 | 394 | 89 | |
| AK171591.1 | - | 3 - 4 | 281 | 89 | ||
| BC031706.1 | - | 3 - 4 | 373 | 4194266 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d23
Genomic Map