Assay Details
| Gene Symbol: Babam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026636.2 | NP_080912.2 | 4 - 5 | 692 | 76 | |
| GenBank mRNA | AK077402.1 | - | 4 - 5 | 678 | 76 | |
| AK147083.1 | - | 4 - 5 | 687 | 76 | ||
| BC005692.1 | - | 4 - 5 | 679 | 3597632 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Babam1
Genomic Map