Assay Details
| Gene Symbol: Babam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026636.2 | NP_080912.2 | 5 - 6 | 770 | 80 | |
| GenBank mRNA | AK077402.1 | - | 5 - 6 | 756 | 80 | |
| AK147083.1 | - | 5 - 6 | 765 | 80 | ||
| AK183167.1 | - | 1 - 2 | 53 | 80 | ||
| BC005692.1 | - | 5 - 6 | 757 | 3597632 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Babam1
Genomic Map