Assay Details
| Gene Symbol: Babam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026636.2 | NP_080912.2 | 7 - 8 | 926 | 90 | |
| GenBank mRNA | AK077402.1 | - | 7 - 8 | 912 | 90 | |
| AK147083.1 | - | 7 - 8 | 921 | 90 | ||
| AK210692.1 | - | 3 - 4 | 184 | 90 | ||
| BC005692.1 | - | 7 - 8 | 913 | 3597632 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Babam1
Genomic Map