Assay Details
| Gene Symbol: Taf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026836.2 | NP_081112.1 | 3 - 4 | 459 | 69 | |
| GenBank mRNA | AK004168.1 | - | 2 - 3 | 106 | 69 | |
| AK050689.1 | - | 3 - 4 | 460 | 69 | ||
| AK133851.1 | - | 4 - 5 | 1443 | 69 | ||
| BC005603.1 | - | 3 - 4 | 438 | 3483315 | 69 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Taf11
Genomic Map