| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100019681.mm.1 |
NM_001164224.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019681.mm.1 |
NM_001164224.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019681.mm.1 |
NM_026877.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019681.mm.1 |
NM_026877.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019681.mm.1 |
NM_198223.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019681.mm.1 |
NM_198223.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019681.mm.1 |
TC1100002043.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100019678.mm.1 |
NM_001164224.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019678.mm.1 |
NM_001164224.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019678.mm.1 |
NM_026877.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019678.mm.1 |
NM_026877.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019678.mm.1 |
NM_198223.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019678.mm.1 |
NM_198223.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019678.mm.1 |
TC1100002043.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100019680.mm.1 |
NM_001164224.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019680.mm.1 |
NM_001164224.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019680.mm.1 |
NM_026877.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019680.mm.1 |
NM_026877.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019680.mm.1 |
NM_198223.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019680.mm.1 |
NM_198223.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019680.mm.1 |
TC1100002043.mm.1 |
MTA TC |