Assay Details
| Gene Symbol: Chmp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026879.2 | NP_081155.1 | 3 - 4 | 506 | 60 | |
| GenBank mRNA | AK084111.1 | - | 3 - 4 | 508 | 60 | |
| BC042626.1 | - | 2 - 3 | 262 | 60 | ||
| BC055809.1 | - | 3 - 4 | 535 | 6414077 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chmp2b
Genomic Map