Assay Details
| Gene Symbol: Ctc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013256.2 | NP_001013274.2 | 16 - 17 | 2965 | 93 | |
| NM_001143790.1 | NP_001137262.1 | 16 - 17 | 2962 | 93 | ||
| NM_001281464.1 | NP_001268393.1 | 16 - 17 | 2833 | 93 | ||
| NM_001281465.1 | NP_001268394.1 | 16 - 17 | 2949 | 93 | ||
| GenBank mRNA | AK038651.1 | - | 16 - 17 | 2749 | 93 | |
| AK044055.1 | - | 16 - 17 | 2744 | 93 | ||
| AK145919.1 | - | 16 - 17 | 2780 | 93 | ||
| AK165906.1 | - | 16 - 17 | 2765 | 93 | ||
| AK198189.1 | - | 2 - 3 | 238 | 93 | ||
| BC014687.1 | - | 7 - 8 | 1133 | 93 | ||
| BC060629.1 | - | 12 - 13 | 2015 | 6847619 | 93 | |
| BC066078.1 | - | 16 - 17 | 2776 | 93 | ||
| BC072625.1 | - | 16 - 17 | 2790 | 93 | ||
| BC147495.1 | - | 16 - 17 | 2763 | 9056325 | 93 | |
| BC147497.1 | - | 16 - 17 | 2763 | 9056354 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctc1
Genomic Map