Assay Details
| Gene Symbol: Ctc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013256.2 | NP_001013274.2 | 19 - 20 | 3366 | 71 | |
| NM_001143790.1 | NP_001137262.1 | 19 - 20 | 3363 | 71 | ||
| NM_001281464.1 | NP_001268393.1 | 19 - 20 | 3234 | 71 | ||
| NM_001281465.1 | NP_001268394.1 | 19 - 20 | 3350 | 71 | ||
| GenBank mRNA | AK038651.1 | - | 19 - 20 | 3150 | 71 | |
| AK044055.1 | - | 19 - 20 | 3145 | 71 | ||
| AK145919.1 | - | 19 - 20 | 3181 | 71 | ||
| AK165906.1 | - | 19 - 20 | 3166 | 71 | ||
| BC014687.1 | - | 10 - 11 | 1534 | 71 | ||
| BC060629.1 | - | 15 - 16 | 2416 | 6847619 | 71 | |
| BC066078.1 | - | 19 - 20 | 3177 | 71 | ||
| BC072625.1 | - | 19 - 20 | 3191 | 71 | ||
| BC147495.1 | - | 19 - 20 | 3164 | 9056325 | 71 | |
| BC147497.1 | - | 19 - 20 | 3164 | 9056354 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctc1
Genomic Map