Assay Details
| Gene Symbol: Cdk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109626.1 | NP_001103096.1 | 2 - 3 | 1979 | 71 | |
| NM_001109628.1 | NP_001103098.1 | 2 - 3 | 1979 | 71 | ||
| NM_026952.2 | NP_081228.2 | 2 - 3 | 1979 | 71 | ||
| GenBank mRNA | AK049376.1 | - | 2 - 3 | 1625 | 71 | |
| AK129237.1 | - | 2 - 3 | 1118 | 71 | ||
| AK164881.1 | - | 2 - 3 | 1840 | 71 | ||
| AY072295.1 | - | 2 - 3 | 1970 | 71 | ||
| BC116645.1 | - | 2 - 3 | 1962 | 40086381 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdk12
Genomic Map