Assay Details
| Gene Symbol: Ints4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027256.2 | NP_081532.1 | 17 - 18 | 2242 | 95 | |
| GenBank mRNA | AK011676.1 | - | 3 - 4 | 239 | 95 | |
| BC013710.1 | - | 17 - 18 | 2188 | 3156446 | 95 | |
| BC013813.1 | - | 5 - 6 | 496 | 95 | ||
| BC021838.1 | - | 10 - 11 | 1245 | 95 | ||
| BC031145.1 | - | 11 - 12 | 1321 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ints4
Genomic Map