Assay Details
| Gene Symbol: Ints4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027256.2 | NP_081532.1 | 1 - 2 | 196 | 67 | |
| GenBank mRNA | AK088635.1 | - | 1 - 2 | 140 | 67 | |
| AK163383.1 | - | 1 - 2 | 150 | 67 | ||
| BC013710.1 | - | 1 - 2 | 142 | 3156446 | 67 | |
| CA450656.1 | - | 1 - 2 | 66 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK163383.1 transcripts.
Set Membership: |
|

Ints4
Genomic Map