Assay Details
| Gene Symbol: Ints4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027256.2 | NP_081532.1 | 20 - 21 | 2589 | 67 | |
| GenBank mRNA | AK011676.1 | - | 6 - 7 | 586 | 67 | |
| BC013710.1 | - | 20 - 21 | 2535 | 3156446 | 67 | |
| BC013813.1 | - | 8 - 9 | 843 | 67 | ||
| BC021838.1 | - | 13 - 14 | 1592 | 67 | ||
| BC031145.1 | - | 14 - 15 | 1668 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ints4
Genomic Map