Assay Details
| Gene Symbol: Ints4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027256.2 | NP_081532.1 | 6 - 7 | 855 | 69 | |
| GenBank mRNA | AK163383.1 | - | 6 - 7 | 811 | 69 | |
| BC013710.1 | - | 6 - 7 | 801 | 3156446 | 69 | |
| CX565861.1 | - | 3 - 4 | 339 | 69 | ||
More Information
Set Membership: |
|

Ints4
Genomic Map