Assay Details
| Gene Symbol: Taf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027427.2 | NP_081703.1 | 5 - 6 | 384 | 67 | |
| GenBank mRNA | AK011843.1 | - | 5 - 6 | 406 | 67 | |
| AK051568.1 | - | 5 - 6 | 385 | 67 | ||
| AK078205.1 | - | 5 - 6 | 396 | 67 | ||
| AK146331.1 | - | 5 - 6 | 347 | 67 | ||
| AK166550.1 | - | 5 - 6 | 377 | 67 | ||
| BC137591.1 | - | 5 - 6 | 358 | 8860610 | 67 | |
More Information
Set Membership: |
|

Taf15
Genomic Map