Assay Details
| Gene Symbol: Cwf19l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027545.2 | NP_081821.1 | 10 - 11 | 1604 | 95 | |
| GenBank mRNA | AK033457.1 | - | 3 - 4 | 348 | 95 | |
| AK037905.1 | - | 2 - 3 | 666 | 95 | ||
| AK045423.1 | - | 10 - 11 | 1605 | 95 | ||
| AK083386.1 | - | 10 - 11 | 1617 | 95 | ||
| AK167727.1 | - | 3 - 4 | 327 | 95 | ||
| BC043480.1 | - | 10 - 11 | 1571 | 95 | ||
| BC125483.1 | - | 10 - 11 | 1641 | 40129998 | 95 | |
| BC125485.1 | - | 10 - 11 | 1641 | 40130000 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cwf19l2
Genomic Map