Assay Details
| Gene Symbol: Tbc1d9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111304.1 | NP_001104774.1 | 18 - 19 | 3626 | 61 | |
| NM_027758.4 | NP_082034.1 | 15 - 16 | 2927 | 61 | ||
| GenBank mRNA | AK017015.1 | - | 4 - 5 | 445 | 61 | |
| AK049049.1 | - | 15 - 16 | 2928 | 61 | ||
| AK077081.1 | - | 4 - 5 | 459 | 61 | ||
| AK134614.1 | - | 18 - 19 | 2903 | 61 | ||
| AK173050.1 | - | 18 - 19 | 2799 | 61 | ||
| BC057027.1 | - | 14 - 15 | 2214 | 6825869 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d9
Genomic Map