| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800006761.mm.1 |
NM_001111304.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006761.mm.1 |
NM_001111304.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006761.mm.1 |
TC0800000916.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006759.mm.1 |
NM_001111304.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006759.mm.1 |
NM_001111304.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006759.mm.1 |
NM_027758.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006759.mm.1 |
NM_027758.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006759.mm.1 |
TC0800000916.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006760.mm.1 |
NM_001111304.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006760.mm.1 |
NM_001111304.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006760.mm.1 |
TC0800000916.mm.1 |
MTA TC |