Assay Details
| Gene Symbol: Slc16a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114332.1 | NP_001107804.1 | 1 - 2 | 551 | 74 | |
| NM_028247.4 | NP_082523.1 | 1 - 2 | 551 | 74 | ||
| GenBank mRNA | AK036743.1 | - | 1 - 2 | 552 | 74 | |
| AK050229.1 | - | 1 - 2 | 554 | 74 | ||
| AK144135.1 | - | 1 - 2 | 554 | 74 | ||
| AK151290.1 | - | 1 - 2 | 552 | 74 | ||
| AK151636.1 | - | 1 - 2 | 552 | 74 | ||
| AK151704.1 | - | 1 - 2 | 556 | 74 | ||
| AK157355.1 | - | 1 - 2 | 540 | 74 | ||
| AK165240.1 | - | 1 - 2 | 552 | 74 | ||
| AK165579.1 | - | 1 - 2 | 554 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc16a10
Genomic Map