Assay Details
| Gene Symbol: Gpbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122963.1 | NP_001116435.1 | 7 - 8 | 1985 | 125 | |
| NM_028487.4 | NP_082763.3 | 8 - 9 | 2045 | 125 | ||
| GenBank mRNA | AK082704.1 | - | 7 - 8 | 1803 | 125 | |
| AK135728.1 | - | 8 - 9 | 1976 | 125 | ||
| AK145318.1 | - | 7 - 8 | 1974 | 125 | ||
| AK151283.1 | - | 6 - 7 | 1206 | 125 | ||
| AK163370.1 | - | 8 - 9 | 1893 | 125 | ||
| AK163390.1 | - | 8 - 9 | 2047 | 125 | ||
| AK169406.1 | - | 8 - 9 | 1840 | 125 | ||
| AY382529.1 | - | 8 - 9 | 1843 | 125 | ||
| BC016083.1 | - | 6 - 7 | 839 | 4501399 | 125 | |
| BC067075.1 | - | 7 - 8 | 1759 | 30614988 | 125 | |
| BC076635.1 | - | 7 - 8 | 1815 | 125 | ||
| BC094888.1 | - | 7 - 8 | 1778 | 30628044 | 125 | |
| BC096442.1 | - | 8 - 9 | 1860 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpbp1
Genomic Map