Assay Details
Gene Symbol: Rbm19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028762.1 | NP_083038.1 | 16 - 17 | 2151 | 87 | |
GenBank mRNA | AK004657.1 | - | 7 - 8 | 966 | 87 | |
AK146493.1 | - | 16 - 17 | 2184 | 87 | ||
AK167100.1 | - | 16 - 17 | 2148 | 87 | ||
BC025619.1 | - | 16 - 17 | 2151 | 5320947 | 87 | |
BC034010.1 | - | 16 - 17 | 2117 | 4225239 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
