Assay Details
| Gene Symbol: Morc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193309.1 | NP_001180238.1 | 6 - 7 | 1092 | 105 | |
| NM_029413.4 | NP_083689.2 | 6 - 7 | 1092 | 105 | ||
| GenBank mRNA | AK017472.1 | - | 6 - 7 | 1061 | 105 | |
| AK032807.1 | - | 6 - 7 | 1093 | 105 | ||
| BC098483.1 | - | 6 - 7 | 1047 | 30022418 | 105 | |
| BC132153.1 | - | 6 - 7 | 946 | 40130430 | 105 | |
| BC132497.1 | - | 6 - 7 | 946 | 40130774 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Morc4
Genomic Map