Assay Details
| Gene Symbol: Golga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290649.1 | NP_001277578.1 | 11 - 12 | 1330 | 90 | |
| NM_029793.2 | NP_084069.1 | 12 - 13 | 1405 | 90 | ||
| NR_110968.1 | 12 - 13 | 1405 | 90 | |||
| GenBank mRNA | AK028516.1 | - | 12 - 13 | 1325 | 90 | |
| AK044987.1 | - | 11 - 12 | 1815 | 90 | ||
| BC037037.1 | - | 7 - 8 | 741 | 90 | ||
| BC043090.1 | - | 12 - 13 | 1270 | 6418353 | 90 | |
| BC053068.1 | - | 11 - 12 | 1258 | 6405595 | 90 | |
| BC080733.1 | - | 11 - 12 | 1330 | 30110235 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Golga1
Genomic Map