Assay Details
| Gene Symbol: Zfp449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030139.4 | NP_084415.1 | 1 - 2 | 43 | 94 | |
| GenBank mRNA | AK036596.1 | - | 1 - 2 | 44 | 94 | |
| AK142888.1 | - | 1 - 2 | 46 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK036596.1, NM_030139.4 transcripts.
Set Membership: |
|

Zfp449
Genomic Map